missing translation for 'onlineSavingsMsg'
Learn More

GFAP Antibody (012), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18334631 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334631 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334631 Supplier Novus Biologicals Supplier No. NBP289203AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GFAP Monoclonal antibody specifically detects GFAP in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFAP
Applications Western Blot
Classification Monoclonal
Clone 12
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias FLJ45472, GFAP astrocytes, glial fibrillary acidic protein
Host Species Rabbit
Immunogen This GFAP Antibody (012) was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the human GFAP.
Molecular Weight of Antigen 50 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2670
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.