missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GF 109203X
GREENER_CHOICE

Product Code. 18025692
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 18025692

Brand: Tocris Bioscience™ 0741/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protein kinase C inhibitor

Very potent and selective inhibitor of protein kinase C, selective for the α and β1 isoforms (IC50 values are 0.0084, 0.0180, 0.210, 0.132, and 5.8 μM for α, β1, δ, ε and ζ isoforms respectively). Selective over MLCK, PKG and PKA (IC50 values are 0.6, 4.6, and 33 μM respectively). Potent antagonist at the 5-HT3 receptor (Ki = 29.5 nM). Anti-inflammatory in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133052-90-1
Molecular Formula C25H24N4O2

Specifications

Quantity 1 mg
Formula Weight 412.49
Purity >98%
Molecular Weight (g/mol) 412.493
Chemical Name or Material GF 109203X
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.