Learn More
GET4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-41084
This item is not returnable.
View return policy
Description
GET4 Polyclonal specifically detects GET4 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| GET4 | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| 0.1 mL |
| Immunoprecipitation | |
| Affinity Purified |