missing translation for 'onlineSavingsMsg'
Learn More

Gestodene Sulfate Pyridinium Salt (>85%), TRC

Product Code. 30391987
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391987 1 mg 1mg
30400900 10 mg 10mg
30405285 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391987 Supplier Toronto Research Chemicals Supplier No. TRCG9425201MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gestodene Sulfate Pyridinium Salt
Analyte or Component Names Gestodene Sulfate Pyridinium Salt
Shipping Condition Blue Ice Dry Pack
Molecular Formula C21 H25 O5 S . C5 H6 N
Synonym (8R,9S,10R,13S,14S,17R)-13-Ethyl-17-ethynyl-3-oxo-2,3,6,7,8,9,10,11,12,13,14,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl Sulfate Pyridin-1-ium
InChI Formula InChI=1S/C21H26O5S.C5H5N/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,4-2)26-27(23,24)25;1-2-4-6-5-3-1/h2,10,12-13,16-19H,3,5-9,11H2,1H3,(H,23,24,25);1-5H/t16-,17+,18+,19-,20-,21-;/m0./s1
Solution Type Neat
SMILES CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1C=C[C@@]2(OS(=O)(=O)[O-])C#C.c5cc[nH+]cc5
IUPAC Name [(8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-3-oxo-1,2,6,7,8,9,10,11,12,14-decahydrocyclopenta[a]phenanthren-17-yl] sulfate;pyridin-1-ium
Industry Type Pharmaceutical
Molecular Weight (g/mol) 469.593
Quantity 1 mg
Formula Weight 469.192
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.