missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gestodene, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Gestodene
Specifications
Specifications
| Chemical Name or Material | Gestodene |
| CAS | 60282-87-3 |
| Analyte or Component Names | Gestodene |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Gestodene, 13-Ethyl-17-hydroxy-18,19-dinor-17a-pregna-4,15-dien-20-yn-3-one, Gestinol, SHB 331, WL 70, 18,19-Dinorpregna-4,15-dien-20-yn-3-one, 13-ethyl-17-hydroxy-, (17alpha)- |
| InChI Formula | InChI=1S/C21H26O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,10,12-13,16-19,23H,3,5-9,11H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1 |
| SMILES | CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1C=C[C@@]2(O)C#C |
| IUPAC Name | (8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-1,2,6,7,8,9,10,11,12,14-decahydrocyclopenta[a]phenanthren-3-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction