missing translation for 'onlineSavingsMsg'
Learn More

Gestodene-3-ethoxy Dienol Ether, TRC

Product Code. 30395378
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395378 5 mg 5mg
30394063 25 mg 25mg
30389294 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395378 Supplier Toronto Research Chemicals Supplier No. TRCG9425155MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gestodene-3-ethoxy Dienol Ether
Analyte or Component Names Gestodene-3-ethoxy Dienol Ether
Shipping Condition Room Temperature
Molecular Formula C23 H30 O2
InChI Formula InChI=1S/C23H30O2/c1-4-22-13-11-19-18-10-8-17(25-6-3)15-16(18)7-9-20(19)21(22)12-14-23(22,24)5-2/h2,7,12,14-15,18-21,24H,4,6,8-11,13H2,1,3H3/t18-,19+,20+,21-,22-,23-/m0/s1
Solution Type Neat
SMILES CCOC1=CC2=CC[C@@H]3[C@H](CC[C@@]4(CC)[C@H]3C=C[C@@]4(O)C#C)[C@H]2CC1
IUPAC Name (8R,9S,10R,13S,14S,17R)-3-ethoxy-13-ethyl-17-ethynyl-2,7,8,9,10,11,12,14-octahydro-1H-cyclopenta[a]phenanthren-17-ol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 338.483
Quantity 5 mg
Formula Weight 338.225
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.