missing translation for 'onlineSavingsMsg'
Learn More

Germanium Standard: Ge @ 1000 μg/mL in H2O, tr. F-

Product Code. 30326445
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326445 100 mL -
30319785 500 mL 500mL
30318020 50 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30326445 Supplier VHG Labs Supplier No. VHGPGEW100
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Germanium
CAS 7440-56-4
Analyte or Component Names Germanium
Shipping Condition Room Temperature
Molecular Formula Ge
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym Ge
InChI Formula InChI=1 S/Ge
Solution Type Single Solution
SMILES [Ge]
Element Ge
IUPAC Name germanium
Solvent or Matrix Water
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 72.64
Quantity 100 mL
Formula Weight 73.9212
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.