missing translation for 'onlineSavingsMsg'
Learn More

Germanium pieces, 3.2mm (0.125in) and down, 99.999% (metals basis)

Product Code. p-4408098
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11385038

Brand: Thermo Scientific Alfa Aesar 043986.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-56-4 | Ge | 72.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-56-4
Molecular Formula Ge
Molecular Weight (g/mol) 72.63
MDL Number MFCD00085310
InChI Key GNPVGFCGXDBREM-UHFFFAOYSA-N
Synonym element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
PubChem CID 6326954
ChEBI CHEBI:30441
IUPAC Name germanium
SMILES [Ge]

Specifications

Color Silver
Physical Form Pieces
Quantity 10 g
Assay Percent Range 99.9999% (Metals basis)
Odor Odorless
Appearance Silver-colored pieces
Vapor Pressure 0 hPa
Density 5.32g/cm3
Chemical Name or Material Germanium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.