missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)

Product Code. 11336178
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336178 25 g 25g
11346178 5 g 5g
11356178 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11336178 Supplier Thermo Scientific Alfa Aesar Supplier No. 011155.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1310-53-8 | GeO2 | 104.628 g/mol

Germanium dioxide is used as an optical material for wide-angle lenses and in optical microscope objective lenses. It is used as a catalyst in the preparation of polyethylene terephthalate resin. Further, it is used as a feedstock for the manufacturing of phosphors and semiconductor materials. It is also used in transistors and diodes, and infrared transmitting glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-53-8
Molecular Formula GeO2
Molecular Weight (g/mol) 104.628
MDL Number MFCD00011030
InChI Key YBMRDBCBODYGJE-UHFFFAOYSA-N
Synonym germanium dioxide, germania, germanic acid, germanium oxide, germanium iv oxide, germanic oxide crystalline, dioxogermanium, germanic oxide, wln: ge o2
PubChem CID 14796
IUPAC Name dioxogermane
SMILES O=[Ge]=O

Specifications

Melting Point 1086°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Slightly soluble in sodium hydroxide. Insoluble in water and hydrochloric acid.Soluble in water,acid and alkali. Insoluble in hydrofluoric acid and hydrochloric acid.
Formula Weight 104.59
Odor Odorless
Density 6.239
Chemical Name or Material Germanium(IV) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.