missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) methoxide, 98%

Product Code. 11488357
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11488357

Brand: Thermo Scientific Alfa Aesar A18437.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 992-91-6 | C4H12GeO4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 992-91-6
Molecular Formula C4H12GeO4
MDL Number MFCD00014881
Synonym tetramethoxygermanium, tetramethoxygermanium iv, methyl germanate iv

Specifications

Boiling Point 145°C
Melting Point -18°C
Quantity 5 g
Linear Formula (CH3O)4Ge
UN Number UN2929
Flash Point 40°C (105°F)
Formula Weight 196.73
Percent Purity 98%
Odor Like Alcohol
Refractive Index 1.4015
Sensitivity Moisture sensitive
Density 1.3254^2^0
Chemical Name or Material Germanium(IV) methoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.