missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) isopropoxide, 99.9% (metals basis)

Product Code. 11365397
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365397 5 g 5g
11345397 1 g 1g
11375397 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11365397 Supplier Thermo Scientific Alfa Aesar Supplier No. 042673.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21154-48-3 | C12H28GeO4 | 308.982 g/mol

Germanium(IV) isopropoxide is used as precursor to prepare GeO2 organic silanes, which is used to study the application of optical wave guide. It is used in the synthesis of heterotactic polyactide and also involved in the macromolecule mediated synthesis of amorphous Germania.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21154-48-3
Molecular Formula C12H28GeO4
Molecular Weight (g/mol) 308.982
MDL Number MFCD00015339
InChI Key PKLMYPSYVKAPOX-UHFFFAOYSA-N
Synonym germanium iv isopropoxide, tetraisopropoxygermane, germanium tetrapropan-2-olate, tetraisopropoxy-germane, tetraisopropoxygermanium, isopropyl germanate iv, germanium tetraisopropoxide
PubChem CID 16685043
IUPAC Name tetra(propan-2-yloxy)germane
SMILES CC(C)O[Ge](OC(C)C)(OC(C)C)OC(C)C

Specifications

Color Colorless
Density 1.025 g/mL
Boiling Point 109°C (30 mmHg)
Flash Point 68°C (154°F)
Odor Alcohol-like
Linear Formula Ge[OCH(CH3)2]4
Refractive Index 1.414
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 308.94
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Germanium (IV) isopropoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.