missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) iodide, 99.999% (metals basis)

Product Code. 11361539
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361539 10 g 10g
11371539 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361539 Supplier Thermo Scientific Alfa Aesar Supplier No. 014009.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13450-95-8 | GeI4 | 580.248 g/mol

Germanium(IV) iodide is a biochemical for proteomics research. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-95-8
Molecular Formula GeI4
Molecular Weight (g/mol) 580.248
MDL Number MFCD00016120
InChI Key CUDGTZJYMWAJFV-UHFFFAOYSA-N
Synonym germanium tetraiodide, germane, tetraiodo, germanium iv iodide, germanium iodide, acmc-1bq9p, germanium iv iodide trace metals basis
PubChem CID 83479
IUPAC Name tetraiodogermane
SMILES [Ge](I)(I)(I)I

Specifications

Melting Point 146°C
Physical Form Crystalline
Quantity 10 g
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 580.21
Percent Purity 99.999%
Sensitivity Moisture sensitive
Density 4.32 g/mL
Assay (metals basis)
Chemical Name or Material Germanium(IV) iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.