missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) ethoxide, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11334708
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Preparation of GeO2 by CVD; intermediate in sol-gel synthesis

Preparation of Germanium oxide by CVD.Sol-gel intermediate for optical fibers and photosensitive SiO2-BeO2 glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14165-55-0
Molecular Formula C8H20GeO4
MDL Number MFCD00015121
Synonym Germanium tetraethoxide; Tetraethoxygermane

Specifications

Melting Point -72°C
Color Colorless
Density 1.14 g/mL
Boiling Point 185.5°C
Flash Point 49°C (121°F)
Odor Alcohol-like
Linear Formula Ge(OC2H5)4
Refractive Index 1.407
Quantity 1 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 252.84
Percent Purity 99.995%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Germanium (IV) ethoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'