missing translation for 'onlineSavingsMsg'
Learn More

Germanium(II) iodide, 99.99% (metals basis)

Product Code. 11391539
Change view
Click to view available options
Quantity:
0.1 g
1 g
5 g
Unit Size:
0.1g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391539 1 g 1g
11381539 0.1 g 0.1g
11301549 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11391539 Supplier Thermo Scientific Alfa Aesar Supplier No. 014013.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13573-08-5 | GeI2 | 326.439 g/mol

Germanium(II) iodide acts as a reagent that forms stable ionic adducts with carbenes. It is also used in the preparation of allylgermanes by reacting with allylstannanes. Further, it reacts with allyl bromide in the presence of zinc iodide to get allylgermanium(IV), which on reacts with carbonyl compounds to give the corresponding homoallylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13573-08-5
Molecular Formula GeI2
Molecular Weight (g/mol) 326.439
MDL Number MFCD00135537
InChI Key IAGYEMVJHPEPGE-UHFFFAOYSA-N
Synonym germanium diiodide, germanium ii iodide, germanium iodide, diiodogermanylidene, acmc-1c83a, germanium ii iodide trace metals basis
PubChem CID 6327215
IUPAC Name diiodogermanium
SMILES [Ge](I)I

Specifications

Color Yellow to Brown
Quantity 1 g
UN Number UN3260
Solubility Information Soluble in dilute hydrochloric acid.
Formula Weight 326.4
Percent Purity 99.99%
Sensitivity Air and moisture sensitive
Density 5.37 g/mL
Assay (metals basis)
Chemical Name or Material Germanium(II) iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.