Learn More
Specac™ Germanium Golden Gate™ Mk ll ATR Top Plate
Product Code. 15290108
Encompass_Preferred
Change view
Click to view available options
Type:
Top Plate
Product Code. Type
15290108 Top Plate
1 options
Product Code. 15290108 Supplier Specac™ Supplier No. GS10566
Only null left
Add to basket
Add to basket
Germanium Top-Plate for use in the Golden Gate Single Reflection ATR Accessory GS10500. Ideal for strongly absorbing samples.
Specifications
For Use With (Equipment) Golden Gate Single Reflection ATR Accessory GS10500
Type Top Plate