Learn More
Specac™ Germanium Golden Gate™ Mk ll ATR Grooved Anvil Wide Gauge
Product Code. 15240108
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Golden Gate ATR Accessory GS10500
Product Code. For Use With (Equipment)
15240108 Golden Gate ATR Accessory GS10500
1 options
Product Code. 15240108 Supplier Specac™ Supplier No. GS10548
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Spare Grooved anvil wide gauge used in the Golden Gate ATR Accessory GS10500. To study the coating on transformer wire the grooved anvils hold the wire exactly in the middle of the diamond.
Specifications
For Use With (Equipment) Golden Gate ATR Accessory GS10500