missing translation for 'onlineSavingsMsg'
Learn More

Germanium Disulfide, MP Biomedicals

Product Code. 11491306 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491306 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11491306 Supplier MP Biomedicals Supplier No. 0520472801
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 12025-34-2
Quantity 1 g
Molecular Formula GeS2
MDL Number MFCD00016122
Synonym germanium disulfide, germanium sulfide, bis sulfanylidene germane, dithioxogermane, germanic sulfide, germanium iv sulfide
InChI Key YIZVROFXIVWAAZ-UHFFFAOYSA-N
SMILES S=[Ge]=S
IUPAC Name germanedithione
Molecular Weight (g/mol) 136.75
PubChem CID 82816
Formula Weight 136.72
Chemical Name or Material Germanium Disulfide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.