missing translation for 'onlineSavingsMsg'
Learn More

Germanium, AAS standard solution, Specpure™ Ge 1000μg/mL, Thermo Scientific Chemicals

£27.40 - £68.50

Chemical Identifiers

MDL Number MFCD00085310
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11369159
View Documents Promotion Details
Thermo Scientific Alfa Aesar
088067.AE
100 mL
£27.40
100mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11336349
View Documents Promotion Details
Thermo Scientific Alfa Aesar
088067.AP
500 mL
£68.50
500mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00085310

Specifications

Ge in 5% HNO3/tr. HF
UN3264
Liquid
MFCD00085310
Specpure™
Germanium, AAS standard solution, Ge 1000μg/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only