missing translation for 'onlineSavingsMsg'
Learn More

Germanium, 99.999%, (trace metal basis), powder

Product Code. p-222452 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 10299203

Brand: Thermo Scientific Acros 193110010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7440-56-4 | Ge | 72.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-56-4
Molecular Formula Ge
Molecular Weight (g/mol) 72.63
MDL Number MFCD00085310
InChI Key GNPVGFCGXDBREM-UHFFFAOYSA-N
Synonym element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
PubChem CID 6326954
ChEBI CHEBI:30441
IUPAC Name germanium
SMILES [Ge]

Specifications

Boiling Point 2830.0°C
Melting Point 937.0°C
Color Black to Gray
Physical Form Glistening Crystalline Powder
Quantity 1 g
Assay Percent Range 99.999%
Merck Index 15, 4441
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in hcl and diluted alkali hydroxides
Formula Weight 72.59
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 5.35
Density 5.3500g/mL
Chemical Name or Material Germanium, 99.999%
Show More Show Less
compliance-icons
Product Identifier
  • Germanium, powder
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.