missing translation for 'onlineSavingsMsg'
Learn More

Geranylgeranyl Thiolacetate, TRC

Product Code. 30391293
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391293 250 mg 250mg
30389646 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391293 Supplier Toronto Research Chemicals Supplier No. TRCG367625250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Geranylgeranyl Thiolacetate
Analyte or Component Names Geranylgeranyl Thiolacetate
Shipping Condition Blue Ice
Molecular Formula C22 H36 O S
Synonym (all-E)-3,7,11,15-Tetramethyl-2,6,10,14-hexadecatetraene-1-thiolacetate, (2E,6E,10E)-3,7,11,15-Tetramethyl-2,6,10,14-hexadecatetraene-1-thiolacetate
InChI Formula InChI=1S/C22H36OS/c1-18(2)10-7-11-19(3)12-8-13-20(4)14-9-15-21(5)16-17-24-22(6)23/h10,12,14,16H,7-9,11,13,15,17H2,1-6H3/b19-12+,20-14+,21-16+
Solution Type Neat
SMILES CC(=CCC\C(=C\CC\C(=C\CC\C(=C\CSC(=O)C)\C)\C)\C)C
IUPAC Name S-[(2E,6E,10E)-3,7,11,15-tetramethylhexadeca-2,6,10,14-tetraenyl] ethanethioate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 348.586
Quantity 250 mg
Formula Weight 348.249
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.