missing translation for 'onlineSavingsMsg'
Learn More

Geranylgeranyl MTS, TRC

Product Code. 30395950
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30395950 5 mg 5mg
30407412 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395950 Supplier Toronto Research Chemicals Supplier No. TRCG3675405MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Geranylgeranyl MTS
Analyte or Component Names Geranylgeranyl MTS
Shipping Condition Room Temperature
Molecular Formula C21 H36 O2 S2
Synonym Geranylgeranyl Methanethiolsulfonate
InChI Formula InChI=1S/C21H36O2S2/c1-18(2)10-7-11-19(3)12-8-13-20(4)14-9-15-21(5)16-17-24-25(6,22)23/h10,12,14,16H,7-9,11,13,15,17H2,1-6H3/b19-12+,20-14+,21-16+
Solution Type Neat
SMILES CC(=CCC\C(=C\CC\C(=C\CC\C(=C\CSS(=O)(=O)C)\C)\C)\C)C
IUPAC Name (2E,6E,10E)-3,7,11,15-tetramethyl-1-methylsulfonylsulfanylhexadeca-2,6,10,14-tetraene
Industry Type Pharmaceutical
Molecular Weight (g/mol) 384.639
Quantity 5 mg
Formula Weight 384.216
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.