missing translation for 'onlineSavingsMsg'
Learn More

Geranylgeraniol (Synthetic), TRC

Product Code. 30405521
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405521 500 mg 500mg
30391043 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30405521 Supplier Toronto Research Chemicals Supplier No. TRCG367501500MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Geranylgeraniol (mixture of isomers)
CAS 24034-73-9
Molecular Formula C20 H34 O
Quantity 500 mg
Synonym (2E,6E,10E)-3,7,11,15-Tetramethyl-2,6,10,14-hexadecatetraen-1-ol, (E,E,E)- 3,7,11,15-Tetramethyl-2,6,10,14-hexadecatetraen-1-ol, (E,E,E)-Geranylgeraniol, All-trans-Geranylgeraniol, trans,trans,trans-Geranylgeraniol, trans-Geranylgeraniol
InChI Formula InChI=1S/C20H34O/c1-17(2)9-6-10-18(3)11-7-12-19(4)13-8-14-20(5)15-16-21/h9,11,13,15,21H,6-8,10,12,14,16H2,1-5H3/b18-11+,19-13+,20-15+
SMILES CC(=CCC\C(=C\CC\C(=C\CC\C(=C\CO)\C)\C)\C)C
IUPAC Name (2E,6E,10E)-3,7,11,15-tetramethylhexadeca-2,6,10,14-tetraen-1-ol
Molecular Weight (g/mol) 290.48
Formula Weight 290.261
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.