missing translation for 'onlineSavingsMsg'
Learn More

Geranyl Butyrate, TRC

Product Code. 30393056
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393056 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393056 Supplier Toronto Research Chemicals Supplier No. TRCG254105100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Geranyl Butyrate
CAS 106-29-6
Molecular Formula C14H24O2
Quantity 100 mg
Synonym AG 76, Geranyl Butanoate, Geranyl Butyrate, Geranyl n-Butyrate, NSC 46146, NSC 70177, trans-3,7-Dimethyl-2,6-octadien-1-yl Butyrate
InChI Formula InChI=1S/C14H24O2/c1-5-7-14(15)16-11-10-13(4)9-6-8-12(2)3/h8,10H,5-7,9,11H2,1-4H3/b13-10+
SMILES CCCC(OC/C=C(CC/C=C(C)\C)\C)=O
IUPAC Name (E)-3,7-dimethylocta-2,6-dien-1-yl butyrate
Molecular Weight (g/mol) 224.34
Formula Weight 224.1776
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.