missing translation for 'onlineSavingsMsg'
Learn More

Geranyl Bromide (>80%), TRC

Product Code. 30408092
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408092 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408092 Supplier Toronto Research Chemicals Supplier No. TRCG3672502.5G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Geranyl Bromide
CAS 6138-90-5
Molecular Formula C10H17Br
Quantity 2.5 g
Synonym (E)-1-Bromo-3,7-dimethyl-2,6-octadiene, (2E)-1-Bromo-3,7-dimethylocta-2,6-diene, (2E)-Geranyl Bromide, (E)-1-Bromo-3,7-dimethyl-2,6-octadiene, Geranyl Bromide, trans-Geranyl Bromide
InChI Formula InChI=1S/C10H17Br/c1-9(2)5-4-6-10(3)7-8-11/h5,7H,4,6,8H2,1-3H3/b10-7+
SMILES CC(=CCC\C(=C\CBr)\C)C
Molecular Weight (g/mol) 217.15
Formula Weight 216.0514
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.