missing translation for 'onlineSavingsMsg'
Learn More

Geranyl Bromide (>80%), TRC

Product Code. 30405883
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30405883 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 30405883

Brand: Toronto Research Chemicals TRCG367250250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Geranyl Bromide
CAS 6138-90-5
Molecular Formula C10H17Br
Quantity 250 mg
Synonym (E)-1-Bromo-3,7-dimethyl-2,6-octadiene, (2E)-1-Bromo-3,7-dimethylocta-2,6-diene, (2E)-Geranyl Bromide, (E)-1-Bromo-3,7-dimethyl-2,6-octadiene, Geranyl Bromide, trans-Geranyl Bromide
InChI Formula InChI=1S/C10H17Br/c1-9(2)5-4-6-10(3)7-8-11/h5,7H,4,6,8H2,1-3H3/b10-7+
SMILES CC(=CCC\C(=C\CBr)\C)C
Molecular Weight (g/mol) 217.15
Formula Weight 216.0514
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.