missing translation for 'onlineSavingsMsg'
Learn More

Geranyl acetate, 98%, Thermo Scientific Chemicals

Catalog Number 11468697
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
105-87-3
C12H20O2
196.29
MFCD00015037
HIGQPQRQIQDZMP-DHZHZOJOSA-N
geranyl acetate, geraniol acetate, bay pine oyster oil, acetic acid, geraniol ester, trans-3,7-dimethyl-2,6-octadien-1-yl acetate, geranyl ethanoate, acetic acid, geranyl ester, unii-3w81yg7p9r, geranyl acetate, cis, 3,7-dimethyl-2,6-octadienyl acetate
1549026
CHEBI:5331
[(2E)-3,7-dimethylocta-2,6-dienyl] acetate
CC(C)=CCC\C(C)=C\COC(C)=O
This item is not returnable. View return policy
105-87-3
C12H20O2
196.29
MFCD00015037
HIGQPQRQIQDZMP-DHZHZOJOSA-N
geranyl acetate, geraniol acetate, bay pine oyster oil, acetic acid, geraniol ester, trans-3,7-dimethyl-2,6-octadien-1-yl acetate, geranyl ethanoate, acetic acid, geranyl ester, unii-3w81yg7p9r, geranyl acetate, cis, 3,7-dimethyl-2,6-octadienyl acetate
1549026
CHEBI:5331
[(2E)-3,7-dimethylocta-2,6-dienyl] acetate
CC(C)=CCC\C(C)=C\COC(C)=O

It is used primarily as a component of perfumes for creams and soaps and as a flavoring ingredient. It is used particularly in rose, lavender and geranium formulations where a sweet fruity or citrus aroma is desired. It is found to have has anti-oxidant, anti-inflammatory and anti-proliferative activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used primarily as a component of perfumes for creams and soaps and as a flavoring ingredient. It is used particularly in rose, lavender and geranium formulations where a sweet fruity or citrus aroma is desired. It is found to have has anti-oxidant, anti-inflammatory and anti-proliferative activities.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY
Density 0.91
Boiling Point 137°C to 139°C (25 mm)
Flash Point 104°C (219°F)
Assay Percent Range 0.98
Refractive Index 1.461
Quantity 100 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 196.29
Percent Purity 98%
Chemical Name or Material Geranyl acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'