missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Geranyl acetate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Geranyl Acetate |
| CAS | 105-87-3 |
| Analyte or Component Names | Geranyl Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H20 O2 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2,6-Octadien-1-ol, 3,7-dimethyl-, 1-acetate, (2E)-, 2,6-Octadien-1-ol, 3,7-dimethyl-, acetate, (2E)- (9CI), 2,6-Octadien-1-ol, 3,7-dimethyl-, acetate, (E)- (8CI), Geraniol acetate (6CI), (E)-3,7-Dimethyl-2,6-octadien-1-ol acetate, (E)-3,7-Dimethyl-2,6-octadienyl acetate, 3,7-Dimethyl-2,6-octadien-1-ol acetate, Acetic acid (2E)-3,7-dimethyl-2,6-octadienyl ester, Acetic acid geraniol ester, Bay pine (oyster) oil, Geranyl acetate, Geranyl ethanoate, NSC 2584, trans-1-Acetoxy-3,7-dimethyl-2,6-octadiene, trans-3,7-Dimethyl-2,6-octadien-1-yl acetate, trans-Geranyl acetate, β-Geranyl acetate |
| InChI Formula | InChI=1S/C12H20O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,8H,5,7,9H2,1-4H3/b11-8+ |
| SMILES | CC(=CCC\C(=C\COC(=O)C)\C)C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction