missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Geraniin

Product Code. 16418998
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418998 Supplier MedChemExpress Supplier No. HYN0472
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Geraniin is a TNF-α releasing inhibitor with numerous activities including anticancer, anti-inflammatory, and anti-hyperglycemic activities, with an IC50 of 43 μM.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
Molecular Weight (g/mol) 952.64 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 60976-49-0
Product Type Natural Products
Purity 98.5%
Solubility DMSO : 100 mg/mL (104.97 mM∣Need ultrasonic)
Target TNF Receptor
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.