missing translation for 'onlineSavingsMsg'
Learn More

Gepotidacin S enantiomer, MedChemExpress

Product Code. 16471639
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471639 1 mg 1mg
16409707 5 mg 5mg
16407178 10 mg 10mg
16403546 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16471639 Supplier Medchem Express Supplier No. HY16742A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Gepotidacin S enantiomer is an S enantionmer of gepotidacin.

Gepotidacin S enantiomer is an S enantionmer of gepotidacin.

Specifications

Chemical Name or Material Gepotidacin S enantiomer
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H28N6O3
Quantity 1 mg
Synonym GSK2140944 S enantiomer
SMILES O=C(C=NC(C=C1)=C23)N3[C@@H](CN4CCC(NCC5=CC(CCCO6)=C6C=N5)CC4)CN2C1=O
Molecular Weight (g/mol) 448.52
Formula Weight 448.52
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.