missing translation for 'onlineSavingsMsg'
Learn More

Gepotidacin, MedChemExpress

Product Code. 16499697
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499697 5 mg 5mg
16497168 10 mg 10mg
16461639 1 mg 1mg
16443197 50 mg 50mg
16437166 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16499697 Supplier Medchem Express Supplier No. HY16742
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Gepotidacin (GSK2140944) is a novel triazaacenaphthylene bacterial type II topoisomerase inhibitor.

Gepotidacin (GSK2140944) is a novel triazaacenaphthylene bacterial type II topoisomerase inhibitor.

Specifications

Chemical Name or Material Gepotidacin
CAS 1075236-89-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H28N6O3
Quantity 5 mg
Synonym GSK2140944
Solubility Information DMSO : 7.14 mg/mL (15.92 mM; Need ultrasonic)
SMILES O=C(C=NC(C=C1)=C23)N3[C@H](CN4CCC(NCC5=CC(CCCO6)=C6C=N5)CC4)CN2C1=O
Molecular Weight (g/mol) 448.52
Formula Weight 448.52
Percent Purity 99.29%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.