missing translation for 'onlineSavingsMsg'
Learn More

Gepirone, TRC

Product Code. 30391845
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391845 250 mg
30399087 100 mg
30405616 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391845 Supplier Toronto Research Chemicals Supplier No. TRCG365000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83928-76-1
Molecular Formula C19 H29 N5 O2
Molecular Weight (g/mol) 359.47
Synonym 2,6-Piperidinedione, 4,4-dimethyl-1-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-, 4,4-Dimethyl-1-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-2,6-piperidinedione, Gepirone
IUPAC Name 4,4-dimethyl-1-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]piperidine-2,6-dione
SMILES CC1(C)CC(=O)N(CCCCN2CCN(CC2)c3ncccn3)C(=O)C1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H29N5O2/c1-19(2)14-16(25)24(17(26)15-19)9-4-3-8-22-10-12-23(13-11-22)18-20-6-5-7-21-18/h5-7H,3-4,8-15H2,1-2H3
Formula Weight 359.2321
Chemical Name or Material Gepirone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.