missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gephyrin Monoclonal Antibody (24GB5585)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557413
This item is not returnable.
View return policy
Description
Gephyrin Monoclonal Antibody for Western Blot, Flow
Gephyrin is a neuronal assembly protein that anchors inhibitory neurotransmitter receptors to the postsynaptic cytoskeleton via high affinity binding to a receptor subunit domain and tubulin dimers. In nonneuronal tissues, the encoded protein is also required for molybdenum cofactor biosynthesis. Mutations in this gene may be associated with the neurological condition hyperplexia and also lead to molybdenum cofactor deficiency. Numerous alternatively spliced transcript variants encoding different isoforms have been described; however, the full-length nature of all transcript variants is not currently known.
Specifications
| Gephyrin | |
| Monoclonal | |
| Unconjugated | |
| Gphn | |
| 5730552E08Rik; AI662856; BC027112; C230040D23; Domain E; Domain G; EGK_18299; Geph; gephyrin; gephyrin isoform; Gph; GPHN; GPHRYN; HKPX1; KIAA1385; MOCODC; Molybdopterin adenylyltransferase; Molybdopterin molybdenumtransferase; MPT adenylyltransferase; MPT Mo-transferase; Putative glycine receptor-tubulin linker protein | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10243, 268566 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 24GB5585 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BUV3, Q9NQX3 | |
| Gphn | |
| Recombinant protein of human GPHN (aa 1-736). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction