missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gephyrin/GPHN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-34859
This item is not returnable.
View return policy
Description
Gephyrin/GPHN Polyclonal specifically detects Gephyrin/GPHN in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative).
Specifications
| Gephyrin/GPHN | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| gephyrin, GPHRYN, KIAA1385GPHGEPH | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10243 | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation (Negative) | |
| NP_001019389.1 | |
| GPHN | |
| The immunogen this antibody was made to, maps to a region between residue 686 to 736 of human Gephyrin using the numbering given in entry NP_001019389.1 (GeneID 10243). | |
| 100 μL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Zebrafish, X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Cetartiodactyla, Domestic duck, Green anole, Wild yak and Southern platyfish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction