missing translation for 'onlineSavingsMsg'
Learn More

Gephyrin/GPHN Antibody [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30734500 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734500 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734500 Supplier Novus Biologicals Supplier No. AF7519I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Gephyrin/GPHN Polyclonal antibody specifically detects Gephyrin/GPHN in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gephyrin/GPHN
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias gephyrin, GPHRYN, KIAA1385GPHGEPH
Host Species Sheep
Immunogen E. coli-derived recombinant human Gephyrin/GPHN, Ala2-Ser188, Accession # Q9NQX3
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10243
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.