missing translation for 'onlineSavingsMsg'
Learn More

GENZ-644282, TRC

Product Code. 30390325
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390325 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390325 Supplier Toronto Research Chemicals Supplier No. TRCG3677605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material GENZ-644282
CAS 529488-28-6
Molecular Formula C22 H21 N3 O5
Quantity 5 mg
Synonym 2,3-Dimethoxy-12-[2-(methylamino)ethyl]benzo[c][1,3]benzodioxolo[5,6-h][1,6]naphthyridin-13(12H)-one
InChI Formula InChI=1S/C22H21N3O5/c1-23-4-5-25-21-14-8-19-20(30-11-29-19)9-16(14)24-10-15(21)12-6-17(27-2)18(28-3)7-13(12)22(25)26/h6-10,23H,4-5,11H2,1-3H3
SMILES CNCCN1C(=O)c2cc(OC)c(OC)cc2c3cnc4cc5OCOc5cc4c13
Molecular Weight (g/mol) 407.42
Formula Weight 407.1481
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.