missing translation for 'onlineSavingsMsg'
Learn More

Genz-123346 free base, MedChemExpress

Product Code. 16485308
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485308 10 mg 10mg
16451517 50 mg 50mg
16433326 25 mg 25mg
16418027 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485308 Supplier Medchem Express Supplier No. HY12744

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Genz-123346 (free base) is an inhibitor of GL1 synthase that blocks the conversion of ceramide to GL1; inhibits GM1 with IC50 value of 14 nM.

Genz-123346 (free base) is an inhibitor of GL1 synthase that blocks the conversion of ceramide to GL1

Specifications

Chemical Name or Material Genz-123346 free base
CAS 491833-30-8
Color White
Purity Grade Notes Research
Molecular Formula C24H38N2O4
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (238.91 mM)
SMILES CCCCCCCCC(N[C@H](CN1CCCC1)[C@@H](C2=CC=C(OCCO3)C3=C2)O)=O
Molecular Weight (g/mol) 418.57
Formula Weight 418.57
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.