missing translation for 'onlineSavingsMsg'
Learn More

Gentisuric Acid, TRC

Product Code. 30240492
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240492 50 mg 50mg
30240473 10 mg 10mg
30240800 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240492 Supplier Toronto Research Chemicals Supplier No. TRCG36776350MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25351-24-0
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.17
Synonym Gentisuric Acid
IUPAC Name 2-[(2,5-dihydroxybenzoyl)amino]acetic acid
SMILES OC(=O)CNC(=O)c1cc(O)ccc1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H9NO5/c11-5-1-2-7(12)6(3-5)9(15)10-4-8(13)14/h1-3,11-12H,4H2,(H,10,15)(H,13,14)
Formula Weight 211.0481
Chemical Name or Material Gentisuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.