missing translation for 'onlineSavingsMsg'
Learn More

Gentiopicroside, TRC

Product Code. 30403042
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403042 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403042 Supplier Toronto Research Chemicals Supplier No. TRCG36310050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gentiopicroside
CAS 20831-76-9
Molecular Formula C16 H20 O9
Quantity 50 mg
Synonym 1H,3H-Pyrano[3,4-c]pyran-1-one, 5-ethenyl-6-(β-D-glucopyranosyloxy)-5,6-dihydro-, (5R,6S)-, 1H,3H-Pyrano[3,4-c]pyran-1-one, 5-ethenyl-6-(β-D-glucopyranosyloxy)-5,6-dihydro-, (5R-trans)-, Gentiopicrin (6CI), Gentiopicroside (7CI,8CI), (5R,6S)-5-Ethenyl-6-(β-D-glucopyranosyloxy)-5,6-dihydro-1H,3H-pyrano[3,4-c]pyran-1-one, NSC 606402
InChI Formula InChI=1S/C16H20O9/c1-2-7-8-3-4-22-14(21)9(8)6-23-15(7)25-16-13(20)12(19)11(18)10(5-17)24-16/h2-3,6-7,10-13,15-20H,1,4-5H2/t7-,10-,11-,12+,13-,15+,16+/m1/s1
SMILES OC[C@H]1O[C@@H](O[C@@H]2OC=C3C(=O)OCC=C3[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name (3S,4R)-4-ethenyl-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4,6-dihydro-3H-pyrano[3,4-c]pyran-8-one
Molecular Weight (g/mol) 356.32
Formula Weight 356.1107
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.