missing translation for 'onlineSavingsMsg'
Learn More

Gentamicin sulfate solution-10, MP Biomedicals™

Product Code. 11491822 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mL
5 x 10 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491822 10 mL 10mL
11401832 5 x 10 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491822 Supplier MP Biomedicals Supplier No. 0210503010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1405-41-0
MDL Number MFCD00270181
Synonym Garamycin, Genoptic

Specifications

Type Gentamicin Sulfate
Physical Form Liquid
Color Colorless to Yellow
Quantity 10 mL
Grade USP
Chemical Name or Material Gentamycin Sulfate Solution - 10, Cell Biology and Analysis
compliance-icons compliance-icons
Product Identifier
  • Gentamicin Sulfate Solution-10
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Gentamicin Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.