missing translation for 'onlineSavingsMsg'
Learn More

Gentamicin sulfate, 600 I.U./mg

Product Code. 15435139
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435139 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15435139 Supplier Thermo Scientific Alfa Aesar Supplier No. J62834.06
  • ONLINE EXCLUSIVE PRICE
    £77.60
    £69.65 / 5g
    Save £7.95
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

Gentamicin sulfate, CAS # 1405-41-0, is an aminoglycoside mainly used to inhibit the growth of Gram-negative bacteria. | CAS: 1405-41-0

Gentamycin Sulfate is a broad-spectrum, aminoglycoside antibiotic used for cell culture which inhibits protein synthesis in sensitive organisms. Reported to have activity against gram-negative bacteria, Pseudomonas aeruginosa, gram-positive Staphylococcus aureus and some mycoplasmas. Reported to have low toxicity to mammalian cells and viruses at certain concentrations, useful for long-term tissue culture studies with viruses. This compound is a mixture of Gentamycin c1, c1a and c2.

Research Use only: Not intended for animal or human diagnostic or therapeutic use.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Gentamicin sulfate is a bactericidal aminoglycoside antibiotic which shows activity against Gram-negative bacteria
• It works by inhibiting protein biosynthesis through binding the bacterial ribosome 30S subunit
Applications
• Gentamicin sulfate has been used in studies to determine wound healing due to its antibiotic and antifungal properties
• It has laboratory research applications in the study of a number of clinical conditions, including actinomycotic infection, appendicitis, cystic fibrosis, osteomyelitis, pelvic inflammatory disease, plague, gonorrhea, and tularemia

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-41-0
MDL Number MFCD00270181

Specifications

Type Gentamicin Sulfate
Physical Form Powder
Color White
Concentration 600 IU/mg
Quantity 5 g
Optical Rotation +117° (c=10 in Water)
Merck Index 14,4392
Solubility Information Soluble in water
Chemical Name or Material Gentamicin sulfate
compliance-icons
Product Identifier
  • Gentamycin sulfate, 600 I.U./mg
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Gentamicin, sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.