Learn More
MP Biomedicals™ Gentamicin Sulfate, 50 mg/mL
Shop All MP Biomedicals ProductsDescription
Gentamicin is an aminoglycoside antibiotic complex that inhibits the growth of a wide variety of Gram-positive and Gram-negative microorganisms, including strains resistant to tetracycline, chloramphenicol, kanamycin, and colistin, particularly strains of Pseudomonas, Proteus, Staphylococcus, and Streptococcus. It is effective against mycoplasma as well.
- Used as a selection agent (gentamicin-resistance gene) in molecular biology applications.
- Key applications: cell culture applications, antimicrobial treatment.
Specifications
Specifications
| Physical Form | Liquid |
| pH | 4 to 6 |
| Sterility | Sterile |
| Color | Clear Colorless to Pale Yellow |
| Concentration | 50 mg/mL |
| Quantity | 1 x 10 mL |
| Synonym | Garamycin, Gentamicin sulfate salt, Gentiomycin C |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.