missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Gentamicin sulfate

Product Code. 15433221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433221 1 g 1g
15575136 5 g 5g
15585136 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15433221 Supplier Thermo Scientific Chemicals Supplier No. 455310010
  • ONLINE EXCLUSIVE PRICE
    £78.00
    £70.65 / 1g
    Save £7.35
    9% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Gentamicin sulfate, CAS # 1405-41-0, is an aminoglycoside mainly used to inhibit the growth of Gram-negative bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Gentamicin sulfate is a bactericidal aminoglycoside antibiotic which shows activity against Gram-negative bacteria
  • It works by inhibiting protein biosynthesis through binding the bacterial ribosome 30S subunit

Applications

  • Gentamicin sulfate has been used in studies to determine wound healing due to its antibiotic and antifungal properties
  • It has laboratory research applications in the study of a number of clinical conditions, including actinomycotic infection, appendicitis, cystic fibrosis, osteomyelitis, pelvic inflammatory disease, plague, gonorrhea, and tularemia
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-41-0
Synonym Garamycin, Gentamicin Sulfate

Specifications

Type Gentamicin Sulfate
Melting Point 218.0°C to 237.0°C
Infrared Spectrum Authentic
Water 15% max.
Quantity 1 g
Solubility Information Solubility in water: soluble.
Percent Purity ≥590 IU/mg (Gentamycin)
Chemical Name or Material Gentamicin sulfate
compliance-icons
Product Identifier
  • Gentamicin sulfate
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.