missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HyClone™ Gentamicin Solution
Click to view available options
Quantity:
10 x 10 mL
Unit Size:
Pack of 10
Description
- Active against gram positive and gram negative bacteria and mycoplasma; useful for long-term virus and tissue culture studies
- Used as an antibacterial or antimycobacterial
- Interferes with bacterial protein synthesis by binding to L6 protein of 50S ribosomal subunit
- Common working concentration is 5μg/mL
- Cytotoxic at >300μg/mL
Specifications
Specifications
| Content And Storage | 15° to 3°C |
| Description | Gentamicin Solution, 50mg/mL |
| Antibiotic Spectrum | Broad Spectrum, Gram-Negative and Gram-Positive Bacteria |
| Concentration | 50 mg/mL |
| Form | Solution |
| Quantity | 10 x 10 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction