missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gentamicin Monoclonal Antibody (GB73.3), Invitrogen™

Mouse Monoclonal Antibody

£368.00 - £916.00

Specifications

Antigen Gentamicin
Clone GB73.3
Concentration 1.3 mg/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17890166
View Documents
Invitrogen™
MA5-46053
100 μg
£368.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11508992
View Documents
Invitrogen™
MA110725
500 μg
£477.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17800476
View Documents
Invitrogen™
MA5-46131
1 mg
£916.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MA1-10725 detects Gentamicin. MA1-10725 has been successfully used in ELISA applications. The MA1-10725 immunogen is Gentamicin, conjugated to KLH.

Gentamicin is an aminoglycoside antibiotic, used to treat many types of bacterial infections, particularly those caused by Gram-negative bacteria. However, gentamicin is not used for Neisseria gonorrhoeae, Neisseria meningitidis or Legionella pneumophila bacterial infections (because of the risk of the patient going into shock from lipid A endotoxin found in certain gram negative organisms).
Specifications

Specifications

Gentamicin
1.3 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.4
Gentamicin, conjugated to KLH.
Primary
-20°C, Avoid Freeze/Thaw Cycles
GB73.3
ELISA
Unconjugated
Mouse
Chemical
Gentacycol; Garamycin; Gentavet; Gentamicinum; C21H43N5O7
IgG2a
Protein G
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.