missing translation for 'onlineSavingsMsg'
Learn More

Gentamicin C1 Deuterated Pentaacetate Salt (>90%), TRC

Product Code. 30399212
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399212 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30399212 Supplier Toronto Research Chemicals Supplier No. TRCG3605870.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gentamicin C1 Deuterated Pentaacetate Salt
CAS 287730-69-2
Molecular Formula C31H63N5O17 (for non-Deuterated)
Quantity 0.5 mg
Synonym O-2-Amino-2,3,4,6,7-pentadeoxy-6-(methylamino)-α-D-ribo-heptopyranosyl-(14)-O-[3-deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(16)]-2-deoxy-D-streptamine Pentaacetate Salt Deuterated
InChI Formula InChI=1S/C21H43N5O7/c1-9(25-3)13-6-5-10(22)19(31-13)32-16-11(23)7-12(24)17(14(16)27)33-20-15(28)18(26-4)21(2,29)8-30-20/h9-20,25-29H,5-8,22-24H2,1-4H3/t9?,10-,11+,12-,13+,14+,15-,16-,17+,18-,19-,20-,21+/m1/s1
SMILES CC(NC)[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O[C@@H]3[C@H](O)[C@@H](NC)[C@@](C)(O)CO3)[C@H](N)C[C@@H]2N)[C@H](N)CC1
Molecular Weight (g/mol) 777.86
Formula Weight 477.3162
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.