Learn More
STEMCELL Technologies Genomic DNA Purification Kit
Product Code. 17199901
Encompass_Preferred
Click to view available options
:
250 Preps
Product Code. 17199901
Brand: STEMCELL Technologies 79020
For purification of genomic DNA from cells or tissue
  • The Genomic DNA Purification Kit uses a membrane-based system to quickly purify high-quality genomic DNA that is suitable for downstream applications such as agarose gel electrophoresis, restriction enzyme digestion, and PCR analysis
  • Genomic DNA can be purified from cells in as little as 20 minutes
  • Using a microcentrifuge, up to 20 mg of tissue (mouse tail or animal tissue) or between 1 x 10^4 to 5 x 10^6 cells can be processed per purification
  • NOTE: Genomic DNA purification from tissue requires overnight digestion with Proteinase K Solution (Catalog #79016) prior to purification
Specifications
No. of Reactions 250 Preps