missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Binding Buffer PC

Produktkod. 11992282
missing translation for 'orderingAttributeHoverText'
Quantity:
125 mL
Förpackningsstorlek
125mL
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 11992282

missing translation for 'mfr': Macherey-Nagel™ 740937

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Binding Buffer PC to be used with NucleoSpin Plant II kits.

Includes two optimised, alternative lysis buffers based on the established CTAB and SDS lysis methods. As plants are very heterogenous and contain many different metabolites such as polyphenols, polysaccharides, or acidic components, NucleoSpin™ Plant II offers two different lysis proceudres for optimal processing of various samples. The silica membrane in the NucleoSpin™ Plant II columns is optimised to improve DNA binding. NucleoSpin™ filters ar included for convenient clarification of lysates.NucleoSpin™ Plant II allows processing of up to 100 mg (wet weight) or 20mg (dry weight) starting material. Depending on the individual sample typical yields are in the range from 1μg to 30μg DNA. The eluted DNA is ready to use for subsequent reactions such as PCR*, restriction analysis and others.Kit components: NucleoSpin™ Plant II columns, filters, collecting tubes 2mL, buffers, RNase A.

Silica membrane technology

  • Yield: 1μg to 30μg
  • Sample size: ≤100 mg wet weight ≤20mg dry weight
  • Elution volume: 100μL
  • Binding capacity: 50μg
  • Preparation time: 30 min./prep
  • NucleoSpin filters included for clarification of lysates
  • RNase A included
  • Format: mini spin column
  • Two alternative lysis buffers included for processing of various samples (PL1 uses CTAB lysis method, PL2 uses SDS lysis method)
For isolation of genomic DNA from plant and fungal material.

Specifikationer

Quantity 125 mL
compliance-icons compliance-icons
Produktbeteckning
  • Buffer PC (125 mL)
Signalord
  • Warning
Riskkategorier
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Faroangivelser
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
Kompletterande information
  • MIXTURE LIST-Contains : ethanol, guanidine hydrochloride
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.