missing translation for 'onlineSavingsMsg'
Learn More

Genkwanin, TRC

Product Code. 30135442
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135442 25 mg 25mg
30136369 100 mg 100mg
17964345 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135442 Supplier Toronto Research Chemicals Supplier No. TRCG36000025MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 437-64-9
Molecular Formula C16 H12 O5
Molecular Weight (g/mol) 284.26
Synonym 4H-1-Benzopyran-4-one, 5-hydroxy-2-(4-hydroxyphenyl)-7-methoxy-, Flavone, 4',5-dihydroxy-7-methoxy- (7CI,8CI), Genkwanin (6CI), 4',5-Dihydroxy-7-methoxyflavone, 5,4'-Dihydroxy-7-methoxyflavone, 5-Hydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-1-benzopyran-4-one, 7-Methylapigenin, 7-O-Methylapigenin, Apigenin 7-O-methyl ether, Apigenin 7-methyl ether, Puddumetin
IUPAC Name 5-hydroxy-2-(4-hydroxyphenyl)-7-methoxychromen-4-one
SMILES COc1cc(O)c2C(=O)C=C(Oc2c1)c3ccc(O)cc3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H12O5/c1-20-11-6-12(18)16-13(19)8-14(21-15(16)7-11)9-2-4-10(17)5-3-9/h2-8,17-18H,1H3
Formula Weight 284.0685
Chemical Name or Material Genkwanin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.