missing translation for 'onlineSavingsMsg'
Learn More

Genkwanin, MedChemExpress

Product Code. 16491488
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491488 5 mg 5mg
16459078 10 mg 10mg
16444877 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491488 Supplier Medchem Express Supplier No. HYN0731
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Genkwanin is a major non-glycosylated flavonoid with anti-flammatory activities.

Genkwanin is a major non-glycosylated flavonoid with anti-flammatory activities.

Specifications

Chemical Name or Material Genkwanin
CAS 437-64-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H12O5
Quantity 5 mg
Synonym Puddumetin
Solubility Information DMSO : 5 mg/mL (17.59 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=C(O)C=C2)OC3=CC(OC)=CC(O)=C13
Molecular Weight (g/mol) 284.26
Formula Weight 284.26
Percent Purity 99.27%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.