missing translation for 'onlineSavingsMsg'
Learn More

Genistein, MedChemExpress

Product Code. 16471736
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471736 500 mg 500mg
16406506 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471736 Supplier Medchem Express Supplier No. HY14596
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Genistein, a soy isoflavone, is a multiple tyrosine kinases (e.g., EGFR) inhibitor which acts as a chemotherapeutic agent against different types of cancer, mainly by altering apoptosis, the cell cycle, and angiogenesis and inhibiting metastasis.

Genistein, a soy isoflavone, is a multiple tyrosine kinases (e.g., EGFR) inhibitor which acts as a chemotherapeutic agent against different types of cancer, mainly by altering apoptosis, the cell cycle, and angiogenesis and inhibiting metastasis.

Specifications

Chemical Name or Material Genistein
CAS 446-72-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10O5
Quantity 500 mg
Synonym NPI 031L
Solubility Information DMSO : ≥ 100 mg/mL (370.04 mM)
SMILES OC1=C2C(OC=C(C3=CC=C(O)C=C3)C2=O)=CC(O)=C1
Molecular Weight (g/mol) 270.24
Formula Weight 270.24
Percent Purity 99.15%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.